![Danofloxacin VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/153/921/d75d15d3-2780-4d7e-bcba-e2830e62c7cc/640/d75d15d3-2780-4d7e-bcba-e2830e62c7cc.png)
Danofloxacin
![Danofloxacin VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/153/921/d75d15d3-2780-4d7e-bcba-e2830e62c7cc/640/d75d15d3-2780-4d7e-bcba-e2830e62c7cc.png)
Synonym(s):
Advocin®
Empirical Formula (Hill Notation):
C19H20FN3O3
CAS Number:
Molecular Weight:
357.38
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing