
Damascenone

Synonym(s):
β-Damascenone, 1-(2,6,6-Trimethylcyclohexa-1,3-dien-1-yl)-2-buten-1-one
Empirical Formula (Hill Notation):
C13H18O
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing