
Chlorhexidine diacetate

Synonym(s):
Chlorhexidine diacetate salt
Empirical Formula (Hill Notation):
C22H30Cl2N10 · 2C2H4O2
CAS Number:
Molecular Weight:
625.55
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing