
Boc-L-1,2,3,4-tetrahydronorharman-3-carboxylic acid

Synonym(s):
(S)-2-Boc-1,2,3,4-tetrahydronorharmane-3-carboxylic acid
Empirical Formula (Hill Notation):
C17H20N2O4
CAS Number:
Molecular Weight:
316.35
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing