
Benazepril Related Compound E

Synonym(s):
(3S)-3-Amino-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid monohydrochloride
Empirical Formula (Hill Notation):
C12H14N2O3 · HCl
CAS Number:
Molecular Weight:
270.71
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2