
Alisol B 23-acetate

Synonym(s):
Alisol B acetate, 23-Acetylalismol B, 23-O-Acetylalisol B, Alisol B 23-acetate
Empirical Formula (Hill Notation):
C32H50O5
CAS Number:
Molecular Weight:
514.74
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing