
Acridine Mutagen ICR 191

Synonym(s):
6-Chloro-9-[3-(2-chloroethylamino)propylamino]-2-methoxyacridine dihydrochloride
Empirical Formula (Hill Notation):
C19H21Cl2N3O · 2HCl
Products
Product Category
Brand
Formula Weight
Showing 1-1 of 1
Compare
Product Number
Description
Pricing