Skip to Content
Merck
Acetanisole ≥98%, FCC, FG

Acetanisole

Acetanisole ≥98%, FCC, FG
Synonym(s):
4′-Methoxyacetophenone, 4-Acetylanisole
Linear Formula:
CH3OC6H4COCH3
CAS Number:
Molecular Weight:
150.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing