1-Hydroxypyrene
Synonym(s):
1-Pyrenol
Empirical Formula (Hill Notation):
C16H10O
CAS Number:
Molecular Weight:
218.25
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing