Merck
  • Normalizing Ovulation Rate by Preferential Reduction of Hepato-Visceral Fat in Adolescent Girls With Polycystic Ovary Syndrome.
Normalizing Ovulation Rate by Preferential Reduction of Hepato-Visceral Fat in Adolescent Girls With Polycystic Ovary Syndrome.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine (2017-07-18)
Lourdes Ibáñez, Luis Del Río, Marta Díaz, Giorgia Sebastiani, Óscar J Pozo, Abel López-Bermejo, Francis de Zegher
ABSTRACT
Polycystic ovary syndrome (PCOS) is an increasingly prevalent disorder in adolescent girls, commonly presenting with hirsutism/oligomenorrhea, commonly treated with an oral contraceptive (OC), and commonly followed by oligoanovulatory subfertility. We tested whether an intervention targeting the reduction of hepato-visceral adiposity is followed by a higher ovulation rate than OC treatment. This randomized, open-label, single-center, pilot proof-of-concept study (12 months on treatment, then 12 months off) was performed in adolescent girls with hirsutism and oligomenorrhea (PCOS by National Institutes of Health; no sexual activity; N = 36; mean age 16 years, body mass index 23.5 kg/m SPIOMET was followed by a 2.5-fold higher ovulation rate than OC (p ≤ .001) and by a 6-fold higher normovulatory fraction (71% vs. 12%; p ≤ .001); oligoanovulation risk after SPIOMET was 65% lower (95% confidence interval, 40%-89%) than after OC. Higher post-treatment ovulation rates related to more on-treatment loss of hepatic fat (r Early SPIOMET treatment for PCOS normalized post-treatment ovulation rates more than OC. Focusing PCOS treatment on early reduction of hepato-visceral fat may prevent part of later oligoanovulatory subfertility.