Merck
Orientation dependent interlayer stacking structure in bilayer MoS
Nanoscale (2017-08-25)
Shanshan Wang, Hidetaka Sawada, Christopher S Allen, Angus I Kirkland, Jamie H Warner
ABSTRACT
We have studied the atomic structure of small secondary domains that nucleate on monolayer MoS
Sigma-Aldrich
Sulfur, purum p.a., ≥99% (T)