- Nitric oxide attenuates overexpression of Giα proteins in vascular smooth muscle cells from SHR: Role of ROS and ROS-mediated signaling.
Nitric oxide attenuates overexpression of Giα proteins in vascular smooth muscle cells from SHR: Role of ROS and ROS-mediated signaling.
Vascular smooth muscle cells (VSMC) from spontaneously hypertensive rats (SHR) exhibit decreased levels of nitric oxide (NO) that may be responsible for the overexpression of Giα proteins that has been shown as a contributing factor for the pathogenesis of hypertension in SHR. The present study was undertaken to investigate if increasing the intracellular levels of NO by NO donor S-Nitroso-N-acetyl-DL-penicillamine (SNAP) could attenuate the enhanced expression of Giα proteins in VSMC from SHR and explore the underlying mechanisms responsible for this response. The expression of Giα proteins and phosphorylation of ERK1/2, growth factor receptors and c-Src was determined by Western blotting using specific antibodies. Treatment of VSMC from SHR with SNAP for 24 hrs decreased the enhanced expression of Giα-2 and Giα-3 proteins and hyperproliferation that was not reversed by 1H (1, 2, 4) oxadiazole (4, 3-a) quinoxalin-1-one (ODQ), an inhibitor of soluble guanylyl cyclase, however, PD98059, a MEK inhibitor restored the SNAP-induced decreased expression of Giα proteins towards control levels. In addition, the increased production of superoxide anion, NAD(P)H oxidase activity, overexpression of AT1 receptor, Nox4, p22phox and p47phox proteins, enhanced levels of TBARS and protein carbonyl, increased phosphorylation of PDGF-R, EGF-R, c-Src and ERK1/2 in VSMC from SHR were all decreased to control levels by SNAP treatment. These results suggest that NO decreased the enhanced expression of Giα-2/3 proteins and hyperproliferation of VSMC from SHR by cGMP-independent mechanism and involves ROS and ROS-mediated transactivation of EGF-R/PDGF-R and MAP kinase signaling pathways.