Merck
  • Liquid chromatography-tandem mass spectrometry method for the determination of thiosulfate in human blood and urine as an indicator of hydrogen sulfide poisoning.
Liquid chromatography-tandem mass spectrometry method for the determination of thiosulfate in human blood and urine as an indicator of hydrogen sulfide poisoning.
Legal medicine (Tokyo, Japan) (2017-01-14)
Chikatoshi Maseda, Akira Hayakawa, Katsuhiro Okuda, Masaru Asari, Hiroki Tanaka, Hiromi Yamada, Shigeki Jin, Kie Horioka, Kotaro Matoba, Hiroshi Shiono, Kazuo Matsubara, Keiko Shimizu
ABSTRACT
Being a stable metabolite of hydrogen sulfide, thiosulfate has been utilized as an index for hydrogen sulfide poisoning (HSP). Thiosulfate analysis is mainly performed using gas chromatography/mass spectrometry (GC-MS) due to its high sensitivity and specificity. The GC-MS analysis requires two-step derivatizations of thiosulfate, and the derivative is not stable in solution as it has a disulfide moiety. To resolve this stability issue, we developed a novel analytical method using liquid chromatography-tandem mass spectrometry (LC-MS/MS) for monitoring the pentafluorobenzyl derivative of thiosulfate (the first reaction product of the GC-MS method) in this study. The established method exhibited high reproducibility despite being a more simplified and rapid procedure compare to the GC-MS method. Phenyl 4-hydroxybenzoate was used as an internal standard because 1,3,5-tribromobenzene which had been used in the GC-MS method was not suitable compound for LC-MS/MS with Electrospray ionization (ESI) negative detection. The linear regression of the peak area ratios versus concentrations was fitted over the concentration ranges of 0.5-250μM and 0.25-250μM in blood and urine, respectively. The validation results satisfied the acceptance criteria for intra- and inter-day accuracy and precision. Blood and urine samples from 12 suspected HSP cases were tested using this method. The thiosulfate concentration detected in the sample coincided well with that determined at the scene of each HSP accident.