Merck
  • Does white coat hypertension require treatment over age 80?: Results of the hypertension in the very elderly trial ambulatory blood pressure side project.
Does white coat hypertension require treatment over age 80?: Results of the hypertension in the very elderly trial ambulatory blood pressure side project.
Hypertension (Dallas, Tex. : 1979) (2012-11-23)
Christopher J Bulpitt, Nigel Beckett, Ruth Peters, Jan A Staessen, Ji-Guang Wang, Marius Comsa, Robert H Fagard, Dan Dumitrascu, Vesselka Gergova, Riitta L Antikainen, Elizabeth Cheek, Chakravarthi Rajkumar
ABSTRACT
White coat hypertension is considered to be a benign condition that does not require antihypertensive treatment. Ambulatory blood pressure (ABP) was measured in 284 participants in the Hypertension in the Very Elderly Trial (HYVET), a double-blind randomized trial of indapamide sustained release 1.5 mg±perindopril 2 to 4 mg versus matching placebo in hypertensive subjects (systolic blood pressure 160-199 mm Hg) aged >80 years. ABP recordings (Diasys Integra II) were obtained in 112 participants at baseline and 186 after an average follow-up of 13 months. At baseline, clinic blood pressure (CBP) exceeded the morning ABP by 32/10 mm Hg. Fifty percent of participants fulfilled the established criteria for white coat hypertension. The highest ABP readings were in the morning (average 140/80 mm Hg), the average night-time pressure was low at 124/72 mm Hg, and the average 24-hour blood pressure was 133/77 mm Hg. During follow-up, the systolic/diastolic blood pressure placebo-active differences averaged 6/5 mm Hg for morning ABP, 8/5 mm Hg for 24-hour ABP, and 13/5 mm Hg for CBP. The lowering of blood pressure over 24 hours supports the reduction in blood pressure with indapamide sustained release±perindopril as the explanation for the reduction in total mortality and cardiovascular events observed in the main HYVET study. Because we estimate that 50% had white coat hypertension in the main study, this condition may benefit from treatment in the very elderly.
Product Description
Supelco
Indapamide, analytical standard, for drug analysis
Sigma-Aldrich
Perindopril erbumine