Merck
  • A nanostructured carbon-reinforced polyisobutylene-based thermoplastic elastomer.
A nanostructured carbon-reinforced polyisobutylene-based thermoplastic elastomer.
Biomaterials (2009-12-26)
Judit E Puskas, Elizabeth A Foreman-Orlowski, Goy Teck Lim, Sara E Porosky, Michelle M Evancho-Chapman, Steven P Schmidt, Mirosława El Fray, Marta Piatek, Piotr Prowans, Krystal Lovejoy
ABSTRACT
This paper presents the synthesis and characterization of a polyisobutylene (PIB)-based nanostructured carbon-reinforced thermoplastic elastomer. This thermoplastic elastomer is based on a self-assembling block copolymer having a branched PIB core carrying -OH functional groups at each branch point, flanked by blocks of poly(isobutylene-co-para-methylstyrene). The block copolymer has thermolabile physical crosslinks and can be processed as a plastic, yet retains its rubbery properties at room temperature. The carbon-reinforced thermoplastic elastomer had more than twice the tensile strength of the neat polymer, exceeding the strength of medical grade silicone rubber, while remaining significantly softer. The carbon-reinforced thermoplastic elastomer displayed a high T(g) of 126 degrees C, rendering the material steam-sterilizable. The carbon also acted as a free radical trap, increasing the onset temperature of thermal decomposition in the neat polymer from 256.6 degrees C to 327.7 degrees C. The carbon-reinforced thermoplastic elastomer had the lowest water contact angle at 82 degrees and surface nano-topography. After 180 days of implantation into rabbit soft tissues, the carbon-reinforced thermoplastic elastomer had the thinnest tissue capsule around the microdumbbell specimens, with no eosinophiles present. The material also showed excellent integration into bones.
Sigma-Aldrich
Polyisobutylene, average Mw ~1,000,000, average Mn ~600,000 by GPC/MALLS, average Mv ~1,200,000
Sigma-Aldrich
Polyisobutylene, average Mw ~500,000, average Mn ~200,000 by GPC/MALLS, average Mv ~420,000