- The detection of 20S-hydroxycholesterol in extracts of rat brains and human placenta by a gas chromatograph/mass spectrometry technique.
The detection of 20S-hydroxycholesterol in extracts of rat brains and human placenta by a gas chromatograph/mass spectrometry technique.
The Journal of steroid biochemistry and molecular biology (2003-06-12)
Ying Yong Lin, Margaret Welch, Seymour Lieberman
PMID12798357
ABSTRACT
The presence of 20(S)-hydroxycholesterol in rat brains and human placenta has been established using the gas chromatography/mass spectrometry (GC/MS) select ion monitoring (SIM) technique. Identification was ensured by three criteria: the specific retention time when the compound emerges from the gas chromatogram and the two m/z ions (201 and 461amu) which are characteristic of its mass spectrum. The possible role of 20(S)-hydroxycholesterol in steroid hormone biosynthesis and in other biological processes is discussed.