- Dual-Color Metal-Induced Energy Transfer (MIET) Imaging for Three-Dimensional Reconstruction of Nuclear Envelope Architecture.
Dual-Color Metal-Induced Energy Transfer (MIET) Imaging for Three-Dimensional Reconstruction of Nuclear Envelope Architecture.
Methods in molecular biology (Clifton, N.J.) (2020-07-19)
Alexey I Chizhik, Anna M Chizhik, Daja Ruhlandt, Janine Pfaff, Narain Karedla, Ingo Gregor, Ralph H Kehlenbach, Jörg Enderlein
PMID32681482
ABSTRACT
The nuclear envelope, comprising the inner and the outer nuclear membrane, separates the nucleus from the cytoplasm and plays a key role in cellular functions. Nuclear pore complexes (NPCs) are embedded in the nuclear envelope and control transport of macromolecules between the two compartments. Recently, it has been shown that the axial distance between the inner nuclear membrane and the cytoplasmic side of the NPC can be measured using dual-color metal-induced energy transfer (MIET). This chapter focuses on experimental aspects of this method and discusses the details of data analysis.