Merck
  • The intracellular pathway for the presentation of vitamin B-related antigens by the antigen-presenting molecule MR1.
The intracellular pathway for the presentation of vitamin B-related antigens by the antigen-presenting molecule MR1.
Nature immunology (2016-04-05)
Hamish E G McWilliam, Sidonia B G Eckle, Alex Theodossis, Ligong Liu, Zhenjun Chen, Jacinta M Wubben, David P Fairlie, Richard A Strugnell, Justine D Mintern, James McCluskey, Jamie Rossjohn, Jose A Villadangos
ABSTRACT
The antigen-presenting molecule MR1 presents vitamin B-related antigens (VitB antigens) to mucosal-associated invariant T (MAIT) cells through an uncharacterized pathway. We show that MR1, unlike other antigen-presenting molecules, does not constitutively present self-ligands. In the steady state it accumulates in a ligand-receptive conformation within the endoplasmic reticulum. VitB antigens reach this location and form a Schiff base with MR1, triggering a 'molecular switch' that allows MR1-VitB antigen complexes to traffic to the plasma membrane. These complexes are endocytosed with kinetics independent of the affinity of the MR1-ligand interaction and are degraded intracellularly, although some MR1 molecules acquire new ligands during passage through endosomes and recycle back to the surface. MR1 antigen presentation is characterized by a rapid 'off-on-off' mechanism that is strictly dependent on antigen availability.