Merck
  • Level of N6-Methyladenosine in Peripheral Blood RNA: A Novel Predictive Biomarker for Gastric Cancer.
Level of N6-Methyladenosine in Peripheral Blood RNA: A Novel Predictive Biomarker for Gastric Cancer.
Clinical chemistry (2020-02-11)
Lichen Ge, Nan Zhang, Zhuojia Chen, Jiaxi Song, Yingmin Wu, Zhuoling Li, Feng Chen, Jia Wu, Dandan Li, Jiexin Li, Cheng Wang, Hongsheng Wang, Junjun Wang
ABSTRACT
Dysregulation of N6-methyladenosine (m6A) is associated with various human diseases including cancer. This study aimed to evaluate the level of m6A as a biomarker for gastric cancer (GC) diagnosis. Peripheral blood samples were collected from 100 GC patients, 30 benign gastric disease (BGD) patients, and 75 healthy controls (HCs). Levels of m6A in total RNA and expression of m6A-related proteins were analyzed. The m6A levels in peripheral blood RNA were significantly increased in the GC group compared with those in the BGD or HC groups; moreover, levels increased with the progression and metastasis of GC and decreased in GC patients after surgery. The area under the curve (AUC) for m6A in the GC group was 0.929 (95% CI, 0.88-0.96), which is markedly greater than the AUCs for carcinoembryonic antigen (CEA; 0.694) and carbohydrate antigen 199 (CA199; 0.603). The combination of CEA and CA199 with m6A improved the AUC to 0.955 (95% CI, 0.91-0.98). The expressions of m6A demethylases ALKBH5 and FTO were significantly downregulated in the GC group compared with the HC group. Coculture with GC cells increased the m6A of RNA in promyelocytic (HL-60) and monocytic (THP-1) leukemia cells and nontumorigenic human peripheral blood B lymphocyte cells (PENG-EBV). Furthermore, a xenograft model enhanced the m6A in peripheral blood RNA of mice. Accordingly, expressions of ALKBH5 and FTO were decreased both in vitro and in vivo. Level of m6A in peripheral blood RNA is a promising noninvasive diagnostic biomarker for GC patients.