Merck
  • Experimental investigation of ionic liquid pretreatment of sugarcane bagasse with 1,3-dimethylimadazolium dimethyl phosphate.
Experimental investigation of ionic liquid pretreatment of sugarcane bagasse with 1,3-dimethylimadazolium dimethyl phosphate.
Bioresource technology (2015-03-26)
Samaneh Bahrani, Sona Raeissi, Mohammad Sarshar
ABSTRACT
In this study, an imidazolium-based ionic liquid (IL), 1,3-dimethylimidazolium dimethyl phosphate ([Mmim][DMP]), was applied for pretreating sugarcane bagasse to produce bioethanol. The main goal of this study was to investigate the feasibility of bagasse pretreatment with this IL, and to verify the effect of different operational parameters on the pretreatment process. Results indicated that temperature and duration of IL-pretreatment have optimum values. Within the range investigated, a maximum fermentable sugar conversion of 70.38% was achieved with this IL at 120°C and 120min. The corresponding value was 28.65% for the untreated biomass. The main cause for the observed enhancement in enzymatic hydrolysis was the reduction of cellulose crystallinity in the IL-pretreated biomass, as compared to the untreated sample, because it resulted in higher accessibility of the enzymes to the biomass after pretreatment. Moreover, the results indicated that aqueous [Mmim][DMP] mixtures are not as effective for pretreatment as the pure IL.
Sigma-Aldrich
1,3-Dimethylimidazolium dimethyl phosphate, ≥98.0% (HPLC)
Pricing and availability is not currently available.