Merck
  • Polybrominated diphenyl ethers and organophosphate esters flame retardants in play mats from China and the exposure risks for children.
Polybrominated diphenyl ethers and organophosphate esters flame retardants in play mats from China and the exposure risks for children.
Environment international (2019-12-31)
Bo Peng, Zi-Min Yu, Chen-Chou Wu, Liang-Ying Liu, Lixi Zeng, Eddy Y Zeng
ABSTRACT
A total of 41 play mats made from different raw materials, such as polyethylene (PE), ethylene-vinyl acetate copolymer (EVA), chemical crosslinked polyethylene (XPE), and polyvinyl chloride (PVC), were obtained from Chinese markets and analyzed for flame retardants. Polybrominated diphenyl ethers (PBDEs) and their replacements, organophosphate esters (OPEs), were measured and the associated exposure risks for children were evaluated. The levels (range; median) of OPEs (6.6-7400; 200 ng g-1) were generally 1-2 orders of magnitude higher than those of PBDEs (0.13-72; 13 ng g-1), consistent with the production and usage trends of flame retardants. The concentrations of both PBDEs and OPEs were the lowest in XPE mats (0.13-5.6; 3.3 ng g-1 for PBDEs and 6.6-320; 47 ng g-1 for OPEs) compared to the other three types. Concentration comparison and compositional analysis suggested that PBDEs and OPEs in play mats were most probably from leaching of raw materials, during production, storage, and/or transport. Children's exposure to PBDEs and OPEs from play mats was estimated for three pathways, i.e., dermal contact, inhalation, and hand-to-mouth ingestion. The combined exposure was 5-6 orders of magnitude lower than the established reference dose values, suggesting no obvious health concern regarding the occurrence of PBDEs and OPEs in play mats. Nevertheless, selection of less contaminated, i.e., XPE mats among those under investigation, by consumers is strongly recommended to minimize any potential exposure risk.