Merck
  • Age-Associated Decrease of the Histone Methyltransferase SUV39H1 in HSC Perturbs Heterochromatin and B Lymphoid Differentiation.
Age-Associated Decrease of the Histone Methyltransferase SUV39H1 in HSC Perturbs Heterochromatin and B Lymphoid Differentiation.
Stem cell reports (2016-06-16)
Dounia Djeghloul, Klaudia Kuranda, Isabelle Kuzniak, Daniela Barbieri, Irina Naguibneva, Caroline Choisy, Jean-Christophe Bories, Christine Dosquet, Marika Pla, Valérie Vanneaux, Gérard Socié, Françoise Porteu, David Garrick, Michele Goodhardt
ABSTRACT
The capacity of hematopoietic stem cells (HSC) to generate B lymphocytes declines with age, contributing to impaired immune function in the elderly. Here we show that the histone methyltransferase SUV39H1 plays an important role in human B lymphoid differentiation and that expression of SUV39H1 decreases with age in both human and mouse HSC, leading to a global reduction in H3K9 trimethylation and perturbed heterochromatin function. Further, we demonstrate that SUV39H1 is a target of microRNA miR-125b, a known regulator of HSC function, and that expression of miR-125b increases with age in human HSC. Overexpression of miR-125b and inhibition of SUV39H1 in young HSC induced loss of B cell potential. Conversely, both inhibition of miR-125 and enforced expression of SUV39H1 improved the capacity of HSC from elderly individuals to generate B cells. Our findings highlight the importance of heterochromatin regulation in HSC aging and B lymphopoiesis.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, serum, Upstate®