Merck
  • Enantioselective synthesis of polysubstituted cyclopentanones by organocatalytic double Michael addition reactions.
Enantioselective synthesis of polysubstituted cyclopentanones by organocatalytic double Michael addition reactions.
Organic letters (2010-08-14)
Anqi Ma, Dawei Ma
ABSTRACT
The O-TMS-protected diphenylprolinol-catalyzed cascade double Michael addition reactions of alpha,beta-unsaturated aldehydes with a beta-keto ester bearing a highly electron-deficient olefin unit occur smoothly to afford polysubstituted cyclopentanones. This process allows formation of four contiguous stereocenters in the cyclopentanone ring in one-step with excellent enantioselectivity.
Product Description
Supelco
Cyclopentanone, analytical standard
Sigma-Aldrich
Cyclopentanone, ≥99%, FG
Sigma-Aldrich
Cyclopentanone, ReagentPlus®, ≥99%