
Zolpidem Related Compound A

Synonym(s):
N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide
Empirical Formula (Hill Notation):
C19H21N3O
Molecular Weight:
307.39
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing