
Ziprasidone hydrochloride

Synonym(s):
Ziprasidone hydrochloride monohydrate, 5-[2-[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6-chloro-1,3-dihydro-2H-indol-2-one hydrochloride monohydrate, CP-88059, Geodon
Empirical Formula (Hill Notation):
C21H21ClN4OS · HCl · H2O
CAS Number:
Molecular Weight:
467.41
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing