
Xylobiose

Synonym(s):
Xylobiose, (2R,3R,4R)-2,3,5-Trihydroxy-4-[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxypentanal, 4-O-β-D-Xylopyranosyl-D-xylose
Empirical Formula (Hill Notation):
C10H18O9
CAS Number:
Molecular Weight:
282.24
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing