
S-tert-Butylmercapto-L-cysteine

Synonym(s):
3-(tert-Butyldithio)-L-alanine, S-(tert-Butylthio)-L-cysteine
Empirical Formula (Hill Notation):
C7H15NO2S2
Products
Product Category
Color
Formula Weight
Physical Form
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing