
Schisandrol B

Synonym(s):
Gomisin A
Empirical Formula (Hill Notation):
C23H28O7
CAS Number:
Molecular Weight:
416.46
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing