Skip to Content
Merck
(R)-1-Boc-3-methylpiperazine 97%

(R)-1-Boc-3-methylpiperazine

(R)-1-Boc-3-methylpiperazine 97%
Synonym(s):
tert-Butyl (R)-3-methyl-1-piperazinecarboxylate
Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing