
Pyridoxal 5′-phosphate solution

Empirical Formula (Hill Notation):
C8H10NO6P
CAS Number:
Molecular Weight:
247.14
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing