
Prochloraz Metabolite BTS44595

Synonym(s):
1-Propyl-1-[2-(2,4,6-trichlorophenoxy)ethyl]urea
Empirical Formula (Hill Notation):
C12H15Cl3N2O2
CAS Number:
Molecular Weight:
325.62
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing