
Plastic additive 22

Synonym(s):
Poly(4-hydroxy-2,2,6,6-tetramethyl-1-piperidine ethanol-alt-1,4-butanedioic acid)
Linear Formula:
(C15H29NO6)n
CAS Number:
Products
Manufacturer
Melting Point (°C)
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing