
Omeprazole Related Compound A

Synonym(s):
Omeprazole sulfone, 5-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl}-1H-benzimidazole, Omeprazole sulphone
Empirical Formula (Hill Notation):
C17H19N3O4S
CAS Number:
Molecular Weight:
361.42
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2