
Neamine

Synonym(s):
2-Deoxy-4-O-(2,6-diamino-2,6-dideoxy-α-D-glucopyranosyl)-D-streptamine
Empirical Formula (Hill Notation):
C12H26N4O6
CAS Number:
Molecular Weight:
322.36
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing