
Morphine Related Compound A

Synonym(s):
Morphine N-oxide, Genomorphine
Empirical Formula (Hill Notation):
C17H19NO4
CAS Number:
Molecular Weight:
301.34
Products
Brand
Formula Weight
Agency
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing