
(-)-Hydroxycitric acid lactone

Synonym(s):
(+)-Garcinia acid, (−)-Hydroxycitric acid lactone, (2S,3S)-3-Hydroxy-5-oxotetrahydrofuran-2,3-dicarboxylic acid, Garcinia lactone
Empirical Formula (Hill Notation):
C6H6O7
CAS Number:
Molecular Weight:
190.11
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing