
Imazethapyr

Synonym(s):
2-(4,5-Dihydro-4-isopropyl-4-methyl-5-oxo-1H-imidazol-2-yl)-5-ethyl-3-pyridinecarboxylic acid
Empirical Formula (Hill Notation):
C15H19N3O3
CAS Number:
Molecular Weight:
289.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2