
Galactose

Synonym(s):
D-(+)-Galactose, Galactose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Analyte Suitability
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing