
Fusidic acid for peak identification

Synonym(s):
Fusidic acid
Empirical Formula (Hill Notation):
C31H48O6
CAS Number:
Molecular Weight:
516.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing