Flumorph
Synonym(s):
(EZ)-4-[3-(3,4-Dimethoxyphenyl)-3-(4-fluorophenyl)acryloyl]morpholine
Empirical Formula (Hill Notation):
C21H22FNO4
CAS Number:
Molecular Weight:
371.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2