
Farnesyl acetate

Empirical Formula (Hill Notation):
C17H28O2
CAS Number:
Molecular Weight:
264.40
Products
Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing