
Bitertanol

Synonym(s):
3,3-Dimethyl-1-(4-phenylphenoxy)-1-(1,2,4-triazol-1-yl)butan-2-ol
Empirical Formula (Hill Notation):
C20H23N3O2
CAS Number:
Molecular Weight:
337.42
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing