Skip to Content
Merck
Acesulfame K Pharmaceutical Secondary Standard; Certified Reference Material

Acesulfame K

Acesulfame K Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide potassium salt
Empirical Formula (Hill Notation):
C4H4KNO4S
CAS Number:
Molecular Weight:
201.24
EC Number:

Products

Brand
Formula Weight
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2