Skip to Content
Merck
All Photos(2)

Key Documents

05407

Supelco

trans-Isoferulic acid

analytical standard

Synonym(s):

trans-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid, trans-3-Hydroxy-4-methoxycinnamic acid, Hesperetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

230-236 °C

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(\C=C\C(O)=O)cc1O

InChI

1S/C10H10O4/c1-14-9-4-2-7(6-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+

InChI key

QURCVMIEKCOAJU-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: salvia valeriana

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mattia Gasperotti et al.
Talanta, 128, 221-230 (2014-07-26)
An increasing number of studies have concerned the profiling of polyphenol microbial metabolites, especially in urine or plasma, but only a few have regarded their accurate quantification. This study reports on a new ultra-performance liquid chromatography tandem mass spectrometry method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service