Merck
  • Stem cell factor and mesenchymal and neural stem cell transplantation in a rat model of Huntington's disease.
Stem cell factor and mesenchymal and neural stem cell transplantation in a rat model of Huntington's disease.
Molecular and cellular neurosciences (2007-12-18)
Kadiombo Bantubungi, David Blum, Laetitia Cuvelier, Sabine Wislet-Gendebien, Bernard Rogister, Emmanuel Brouillet, Serge N Schiffmann
ABSTRACT
Neural and mesenchymal stem cells have been proposed as alternative sources of cells for transplantation into the brain in neurodegenerative disorders. However, the endogenous factors controlling their engraftment within the injured parenchyma remain ill-defined. Here, we demonstrate significant engraftment of undifferentiated exogenous mesenchymal or neural stem cells throughout the lesioned area in a rat model for Huntington's disease, as late as 8 weeks post-transplantation. We show that stem cell factor (SCF), strongly up-regulated within host cells in the damaged striatum, is able to activate the SCF receptor c-kit and its signaling pathway and to promote the migration and proliferation of mesenchymal and neural stem cells in vitro. Furthermore, c-kit receptor blockade alters neural stem cell distribution within the lesioned striatum. Host SCF expression is observed in atypical cells expressing glial fibrillary acidic protein and doublecortin in the lesioned striatum and in migrating doublecortin-positive progenitors. Taken together, these data demonstrate that SCF produced in situ in the lesioned striatum is an important factor in promoting the engraftment of stem cells within the lesioned brain.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid