Merck
  • Atomic mass tag of bismuth-209 for increasing the immunoassay multiplexing capacity of mass cytometry.
Atomic mass tag of bismuth-209 for increasing the immunoassay multiplexing capacity of mass cytometry.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2017-12-06)
Guojun Han, Shih-Yu Chen, Veronica D Gonzalez, Eli R Zunder, Wendy J Fantl, Garry P Nolan
ABSTRACT
Mass cytometry (or CyTOF) is an atomic mass spectrometry-based single-cell immunoassay technology, which has provided an increasingly systematic and sophisticated view in basic biological and clinical studies. Using elemental reporters composed of stable heavy metal isotopes, more than 50 cellular parameters are measured simultaneously. However, this current multiplexing does not meet the theoretical capability of CyTOF instrumentation with 135 detectable channels, primarily due to the limitation of available chemistries for conjugating elemental mass tags to affinity reagents. To address this issue, we develop herein additional metallic mass tag based on bismuth-209 (
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline