Merck
  • Selective regulation of clathrin-mediated epidermal growth factor receptor signaling and endocytosis by phospholipase C and calcium.
Selective regulation of clathrin-mediated epidermal growth factor receptor signaling and endocytosis by phospholipase C and calcium.
Molecular biology of the cell (2017-08-18)
Ralph Christian Delos Santos, Stephen Bautista, Stefanie Lucarelli, Leslie N Bone, Roya M Dayam, John Abousawan, Roberto J Botelho, Costin N Antonescu
ABSTRACT
Clathrin-mediated endocytosis is a major regulator of cell-surface protein internalization. Clathrin and other proteins assemble into small invaginating structures at the plasma membrane termed clathrin-coated pits (CCPs) that mediate vesicle formation. In addition, epidermal growth factor receptor (EGFR) signaling is regulated by its accumulation within CCPs. Given the diversity of proteins regulated by clathrin-mediated endocytosis, how this process may distinctly regulate specific receptors is a key question. We examined the selective regulation of clathrin-dependent EGFR signaling and endocytosis. We find that perturbations of phospholipase Cγ1 (PLCγ1), Ca
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Xestospongin C, film