Merck
  • Synthesis of bicyclic p-diiodobenzenes via silver-catalyzed Csp-H iodination and ruthenium-catalyzed cycloaddition.
Synthesis of bicyclic p-diiodobenzenes via silver-catalyzed Csp-H iodination and ruthenium-catalyzed cycloaddition.
Journal of the American Chemical Society (2006-06-22)
Yoshihiko Yamamoto, Kozo Hattori, Hisao Nishiyama
ABSTRACT
Highly substituted iodobenzenes were efficiently and regioselectively synthesized from readily available 1,6-diynes via two-step process consisting of silver-catalyzed Csp-H iodination and subsequent ruthenium-catalyzed [2 + 2 + 2] cycloaddition of resultant iododiynes. Some of the obtained iodobenzenes were subjected to palladium-catalyzed C-C bond-forming reactions such as Mizoroki-Heck reaction, Sonogashira reaction, and Suzuki-Miyaura coupling, giving highly conjugated molecules.
Sigma-Aldrich
N-Iodosuccinimide, 95%
Pricing and availability is not currently available.