Merck
  • Estimation of the net energy requirements for maintenance in growing and finishing pigs.
Estimation of the net energy requirements for maintenance in growing and finishing pigs.
Journal of animal science (2014-05-08)
G F Zhang, D W Liu, F L Wang, D F Li
ABSTRACT
The objective of this experiment was to determine the net energy requirements for maintenance of growing and finishing pigs using regression models. Thirty-six growing (27.38 ± 2.24 kg) and 36 finishing (70.25 ± 2.61 kg) barrows were used and within each phase. Pigs received a corn-soybean meal diet fed at 6 levels of feed intake, which were calculated as 0, 20, 40, 60, 80, or 100% of the estimated ad libitum ME intake (2,400 kJ ME/kg BW(0.6)·d(-1)) of the pigs. Measurements were conducted on 6 pigs per feeding level and per stage of growth. After a 5-d adjustment period, barrows in the fasted treatment were kept in respiration chambers for 2 d to measure the fasting heat production. Barrows in the other treatments were kept individually in respiration chambers for a 5-d balance trial followed by a 2-d fasting period. Heat production (HP) in the fed state was measured and feces and urine were collected in the balance trial. The total HP increased (P < 0.01) with increasing feeding levels. Fasting HP increased (P < 0.01) as previous feeding level increased and was less (P = 0.012) in finishing pigs than growing pigs if calculated per kilogram BW(0.6) per day. When using an exponential regression analysis, ME requirements for maintenance were estimated at 973 and 921 kJ/kg BW(0.6)·d(-1) and NE requirements for maintenance were estimated at 758 and 732 kJ/kg BW(0.6)·d(-1) for growing and finishing pigs, respectively. The efficiencies of using ME for growth and for maintenance were estimated at 66 and 78.7% for growing and finishing pigs, respectively. It is concluded that exponential regression between HP and a wide range of ME intake may be used as a new method to determine the NE requirement for maintenance.
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%