Merck
  • Evidence for an oxygen evolving iron-oxo-cerium intermediate in iron-catalysed water oxidation.
Evidence for an oxygen evolving iron-oxo-cerium intermediate in iron-catalysed water oxidation.
Nature communications (2015-01-23)
Zoel Codolà, Laura Gómez, Scott T Kleespies, Lawrence Que, Miquel Costas, Julio Lloret-Fillol
ABSTRACT
The non-haem iron complex α-[Fe(II)(CF3SO3)2(mcp)] (mcp=(N,N'-dimethyl-N,N'-bis(2-pyridylmethyl)-1,2-cis-diaminocyclohexane) reacts with Ce(IV) to oxidize water to O2, representing an iron-based functional model for the oxygen evolving complex of photosystem II. Here we trap an intermediate, characterized by cryospray ionization high resolution mass spectrometry and resonance Raman spectroscopy, and formulated as [(mcp)Fe(IV)(O)(μ-O)Ce(IV)(NO3)3](+), the first example of a well-characterized inner-sphere complex to be formed in cerium(IV)-mediated water oxidation. The identification of this reactive Fe(IV)-O-Ce(IV) adduct may open new pathways to validate mechanistic notions of an analogous Mn(V)-O-Ca(II) unit in the oxygen evolving complex that is responsible for carrying out the key O-O bond forming step.
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium nitrate, BioXtra, ≥99.5%
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture
Sigma-Aldrich
Ammonium nitrate, ≥99.0%
Sigma-Aldrich
Ammonium nitrate, ACS reagent, ≥98%